Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Athl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145387.4 | NP_663362.2 | 3 - 4 | 551 | 109 | |
GenBank mRNA | AK077662.1 | - | 3 - 4 | 552 | 109 | |
AK085600.1 | - | 3 - 4 | 567 | 109 | ||
AK171694.1 | - | 2 - 3 | 461 | 109 | ||
BC056953.1 | - | 3 - 4 | 513 | 5717810 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |