Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142681.1 | NP_001136153.1 | 7 - 8 | 739 | 63 | |
NM_026522.5 | NP_080798.2 | 7 - 8 | 739 | 63 | ||
GenBank mRNA | AK014071.1 | - | 7 - 8 | 688 | 63 | |
AK050655.1 | - | 7 - 8 | 687 | 63 | ||
AK160156.1 | - | 7 - 8 | 699 | 63 | ||
BC006876.1 | - | 7 - 8 | 688 | 2647540 | 63 | |
BC061063.1 | - | 7 - 8 | 699 | 6591338 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |