Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hirip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172746.3 | NP_766334.1 | 4 - 5 | 1856 | 118 | |
GenBank mRNA | AK045122.1 | - | 4 - 5 | 1391 | 118 | |
AK135526.1 | - | 4 - 5 | 1401 | 118 | ||
AK161657.1 | - | 4 - 5 | 1391 | 118 | ||
AK162096.1 | - | 1 - 2 | 418 | 118 | ||
BC055687.1 | - | 4 - 5 | 1856 | 6414655 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |