Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178641.5 | NP_848756.2 | 8 - 9 | 1279 | 69 | |
GenBank mRNA | AK030870.1 | - | 8 - 9 | 1281 | 69 | |
AK039468.1 | - | 7 - 8 | 1277 | 69 | ||
AK129249.1 | - | 8 - 9 | 1236 | 69 | ||
AK150418.1 | - | 8 - 9 | 1319 | 69 | ||
AK154996.1 | - | 8 - 9 | 1284 | 69 | ||
BC125437.1 | - | 8 - 9 | 1167 | 40129952 | 69 | |
BC137700.1 | - | 8 - 9 | 1167 | 8860719 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |