Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242349.1 | NP_001229278.1 | 14 - 15 | 1683 | 81 | |
NM_178642.5 | NP_848757.4 | 15 - 16 | 1692 | 81 | ||
GenBank mRNA | AK029329.1 | - | 1 - 2 | 59 | 81 | |
AK032793.1 | - | 10 - 11 | 704 | 81 | ||
AK052589.1 | - | 15 - 16 | 1644 | 81 | ||
BC004697.1 | - | 7 - 8 | 558 | 81 | ||
BC062959.1 | - | 14 - 15 | 1569 | 30547439 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |