Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242349.1 | NP_001229278.1 | 3 - 4 | 714 | 61 | |
NM_178642.5 | NP_848757.4 | 3 - 4 | 711 | 61 | ||
GenBank mRNA | AK052589.1 | - | 3 - 4 | 663 | 61 | |
BC062959.1 | - | 3 - 4 | 600 | 30547439 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |