Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199632.1 | NP_001186561.1 | 17 - 18 | 1960 | 125 | |
NM_177076.4 | NP_796050.2 | 16 - 17 | 1861 | 125 | ||
GenBank mRNA | AK029566.1 | - | 16 - 17 | 1863 | 125 | |
AK030172.1 | - | 17 - 18 | 1944 | 125 | ||
BC145291.1 | - | 17 - 18 | 1886 | 9053845 | 125 | |
BC145292.1 | - | 16 - 17 | 1788 | 9053846 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |