Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D10Jhu81e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138601.2 | NP_613067.1 | 3 - 4 | 338 | 84 | |
GenBank mRNA | AK003866.1 | - | 3 - 4 | 339 | 84 | |
BC013475.1 | - | 3 - 4 | 331 | 4237589 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |