Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146023.1 | NP_001139495.1 | 5 - 6 | 651 | 58 | |
GenBank mRNA | AK080790.1 | - | 4 - 5 | 743 | 58 | |
AK138395.1 | - | 5 - 6 | 625 | 58 | ||
AK166139.1 | - | 5 - 6 | 621 | 58 | ||
AK171108.1 | - | 3 - 4 | 1017 | 58 | ||
BC023421.1 | - | 2 - 3 | 269 | 5029298 | 58 | |
BC058765.1 | - | 4 - 5 | 708 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |