Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175450.4 | NP_780659.2 | 6 - 7 | 815 | 64 | |
GenBank mRNA | AK050106.1 | - | 6 - 7 | 813 | 64 | |
AK082695.1 | - | 6 - 7 | 818 | 64 | ||
AK133716.1 | - | 6 - 7 | 816 | 64 | ||
AK160713.1 | - | 6 - 7 | 813 | 64 | ||
AK166113.1 | - | 813 | 64 | |||
AK168691.1 | - | 6 - 7 | 813 | 64 | ||
BC032968.1 | - | 5 - 6 | 507 | 64 | ||
BC095984.1 | - | 6 - 7 | 815 | 4978914 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |