Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142701.1 | NP_001136173.1 | 21 - 22 | 3099 | 84 | |
NM_027521.3 | NP_081797.1 | 20 - 21 | 2651 | 84 | ||
GenBank mRNA | AK018130.1 | - | 8 - 9 | 84 | ||
AK044860.1 | - | 20 - 21 | 2613 | 84 | ||
AK134029.1 | - | 14 - 15 | 2073 | 84 | ||
AK151423.1 | - | 21 - 22 | 3043 | 84 | ||
AK159194.1 | - | 21 - 22 | 3100 | 84 | ||
AK159474.1 | - | 20 - 21 | 3144 | 84 | ||
AK161766.1 | - | 13 - 14 | 2159 | 84 | ||
AK170262.1 | - | 21 - 22 | 3085 | 84 | ||
AK171304.1 | - | 21 - 22 | 3085 | 84 | ||
BC039802.1 | - | 2 - 3 | 308 | 84 | ||
BC053750.1 | - | 20 - 21 | 2651 | 6333971 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |