Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142701.1 | NP_001136173.1 | 7 - 8 | 1082 | 70 | |
NM_027521.3 | NP_081797.1 | 6 - 7 | 634 | 70 | ||
GenBank mRNA | AK044860.1 | - | 6 - 7 | 596 | 70 | |
AK151423.1 | - | 7 - 8 | 1026 | 70 | ||
AK159194.1 | - | 7 - 8 | 1083 | 70 | ||
AK170262.1 | - | 7 - 8 | 1068 | 70 | ||
AK171304.1 | - | 7 - 8 | 1068 | 70 | ||
BC053750.1 | - | 6 - 7 | 634 | 6333971 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |