Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033324.2 | NP_201581.2 | 9 - 10 | 2127 | 82 | |
GenBank mRNA | AB086857.1 | - | 9 - 10 | 2127 | 82 | |
AK086233.1 | - | 8 - 9 | 2319 | 82 | ||
BC051013.1 | - | 7 - 8 | 2313 | 82 | ||
BC062919.1 | - | 9 - 10 | 2147 | 6833280 | 82 | |
BC066118.1 | - | 9 - 10 | 2174 | 6400561 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |