Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034895.3 | NP_001030067.2 | 15 - 16 | 2264 | 117 | |
GenBank mRNA | BC072571.1 | - | 14 - 15 | 2933 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |