Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039519.2 | NP_001034608.1 | 3 - 4 | 339 | 112 | |
NM_001282083.1 | NP_001269012.1 | 3 - 4 | 630 | 112 | ||
GenBank mRNA | AK050621.1 | - | 3 - 4 | 608 | 112 | |
AK169778.1 | - | 3 - 4 | 306 | 112 | ||
BG088284.2 | - | 3 - 4 | 298 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |