Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146003.2 | NP_666115.2 | 9 - 10 | 1901 | 70 | |
GenBank mRNA | AK048485.1 | - | 4 - 5 | 1350 | 70 | |
AK129219.1 | - | 5 - 6 | 1428 | 70 | ||
BC052718.1 | - | 9 - 10 | 1668 | 70 | ||
BC061480.1 | - | 9 - 10 | 1725 | 6509771 | 70 | |
BC151128.1 | - | 10 - 11 | 1332 | 9088041 | 70 | |
BC172171.1 | - | 10 - 11 | 1332 | 9054841 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |