Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053208.4 | NP_444438.2 | 3 - 4 | 1353 | 109 | |
GenBank mRNA | AF340231.1 | - | 2 - 3 | 1095 | 109 | |
AF453879.1 | - | 3 - 4 | 1275 | 109 | ||
AJ310547.1 | - | 3 - 4 | 1324 | 109 | ||
AK075582.1 | - | 3 - 4 | 1305 | 109 | ||
AK152056.1 | - | 3 - 4 | 1314 | 109 | ||
AK152223.1 | - | 3 - 4 | 1311 | 109 | ||
AK159206.1 | - | 3 - 4 | 1306 | 109 | ||
AK166661.1 | - | 4 - 5 | 1219 | 109 | ||
AK167477.1 | - | 3 - 4 | 1306 | 109 | ||
BC023299.1 | - | 3 - 4 | 1287 | 5135579 | 109 | |
BC051436.1 | - | 2 - 3 | 1071 | 6312638 | 109 | |
BC086764.1 | - | 3 - 4 | 1292 | 5685311 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |