Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009163.3 | NP_033189.2 | 9 - 10 | 997 | 84 | |
GenBank mRNA | AF036894.1 | - | 8 - 9 | 867 | 84 | |
AK036747.1 | - | 9 - 10 | 885 | 84 | ||
AK037789.1 | - | 9 - 10 | 882 | 84 | ||
AK043024.1 | - | 9 - 10 | 884 | 84 | ||
AK049342.1 | - | 9 - 10 | 905 | 84 | ||
AK150053.1 | - | 9 - 10 | 892 | 84 | ||
AK173137.1 | - | 9 - 10 | 926 | 84 | ||
BC026135.1 | - | 9 - 10 | 943 | 4212293 | 84 | |
CT010214.1 | - | 8 - 9 | 807 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |