Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbata | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017419.2 | NP_001017419.1 | 8 - 9 | 1126 | 89 | |
NM_001017433.2 | NP_001017433.1 | 8 - 9 | 1228 | 89 | ||
GenBank mRNA | AF521591.1 | - | 7 - 8 | 1057 | 89 | |
AF521592.1 | - | 7 - 8 | 1159 | 89 | ||
AK006213.1 | - | 7 - 8 | 1078 | 89 | ||
AW490515.1 | - | 2 - 3 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |