Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195268.1 | NP_001182197.1 | 6 - 7 | 753 | 92 | |
NR_036582.1 | 8 - 9 | 1089 | 92 | |||
GenBank mRNA | AK137009.1 | - | 8 - 9 | 1089 | 92 | |
BC052075.1 | - | 7 - 8 | 807 | 92 | ||
BC078442.1 | - | 2 - 3 | 308 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |