Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27256923 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27256924 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27256922 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2008747 |
AK160321.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2008747 |
NM_026095.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2008747 |
AK005341.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2008747 |
AK006692.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
640718 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
535005 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
683217 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
532301 |
BC004761.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900729 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
416760 |
AJ487854.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
416760 |
AK220417.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
416760 |
BC145695.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
416760 |
NM_153135.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
595844 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
546478 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838078 |
AF330197.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838078 |
AK036351.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838078 |
AK048110.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838078 |
AK137148.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838078 |
AK142598.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838078 |
AK164502.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838078 |
AK169999.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838078 |
BC011463.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838078 |
BC026485.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838078 |
BC054842.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838078 |
BC069835.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838078 |
NM_033604.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
86573 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
98905 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
89144 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P115468 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P179423 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2008747 |
AK160321.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2008747 |
NM_026095.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2008747 |
AK005341.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2008747 |
AK006692.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256897 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256886 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256885 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256893 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256899 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256909 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256907 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256883 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256928 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256921 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256912 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256923 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256937 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256906 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256905 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256918 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256933 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256917 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256895 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256889 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256891 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256916 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256902 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256892 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256931 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256924 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256887 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256904 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256929 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256903 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256922 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256934 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256913 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256935 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256920 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256910 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256900 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256939 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256884 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27256925 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07110533 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P115468 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02161488 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P179423 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P523179 |
BC011510.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P523179 |
AK160321.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P523179 |
NM_026095.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P523179 |
AK005341.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P523179 |
AK006692.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P150588 |
AK163643.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P176681 |
BC011510.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P176681 |
AK160321.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P176681 |
NM_026095.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P176681 |
AK005341.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P176681 |
AK006692.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P122725 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P179423 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P115468 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08668997 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08669000 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27256923 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27256924 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27256922 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257041 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257017 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256931 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256966 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256939 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257046 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256968 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256922 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256993 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256995 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256916 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256925 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257030 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256976 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256949 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256941 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257013 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256990 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256985 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256945 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257025 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256981 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257001 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256940 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256998 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256920 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256938 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257014 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256937 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256960 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257047 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256918 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256928 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256973 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256933 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256944 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257021 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256961 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257000 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256954 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257029 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256934 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256978 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256950 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257033 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256947 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257037 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256935 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257006 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257031 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257004 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27257007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27256988 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08327098 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256939 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27257025 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08602354 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256968 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256947 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08230197 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08629446 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256933 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08839309 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08587525 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08520133 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256940 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256957 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256954 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256963 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256958 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08764309 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08333102 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256973 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27257029 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08697310 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256974 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08374706 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256935 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256950 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256945 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256969 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27257019 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08668994 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256920 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256925 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256952 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256938 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08669000 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08839307 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08251896 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27257028 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256955 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256944 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256928 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256961 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256970 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27256937 |
- |
- |