Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252658.1 | NP_001239587.1 | 1 - 2 | 267 | 74 | |
NM_010700.3 | NP_034830.2 | 1 - 2 | 267 | 74 | ||
GenBank mRNA | AF425607.1 | - | 1 - 2 | 115 | 74 | |
AK018697.1 | - | 1 - 2 | 203 | 74 | ||
AK149950.1 | - | 1 - 2 | 205 | 74 | ||
AK151968.1 | - | 1 - 2 | 234 | 74 | ||
AK152103.1 | - | 1 - 2 | 203 | 74 | ||
AK160006.1 | - | 1 - 2 | 205 | 74 | ||
AK161104.1 | - | 1 - 2 | 205 | 74 | ||
AK170264.1 | - | 1 - 2 | 212 | 74 | ||
AK184620.1 | - | 1 - 2 | 119 | 74 | ||
BC019207.1 | - | 1 - 2 | 185 | 5052599 | 74 | |
BC053041.1 | - | 1 - 2 | 212 | 5704871 | 74 | |
BY235019.1 | - | 1 - 2 | 249 | 74 | ||
X64414.1 | - | 1 - 2 | 186 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |