Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027823.1 | NP_082099.1 | 9 - 10 | 1175 | 67 | |
GenBank mRNA | AK082857.1 | - | 9 - 10 | 1263 | 67 | |
BC137891.1 | - | 9 - 10 | 1175 | 8860910 | 67 | |
BC137892.1 | - | 9 - 10 | 1175 | 8860911 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |