Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207176.3 | NP_997059.1 | 1 - 2 | 167 | 73 | |
GenBank mRNA | AK132249.1 | - | 1 - 2 | 124 | 73 | |
AK145609.1 | - | 1 - 2 | 116 | 73 | ||
AK167101.1 | - | 1 - 2 | 116 | 73 | ||
AK167274.1 | - | 1 - 2 | 124 | 73 | ||
BC010465.1 | - | 1 - 2 | 123 | 3585670 | 73 | |
BC051103.1 | - | 1 - 2 | 101 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |