Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161483.1 | NP_001154955.1 | 8 - 9 | 1269 | 63 | |
GenBank mRNA | AK086574.1 | - | 4 - 5 | 773 | 63 | |
BC056377.1 | - | 8 - 9 | 1262 | 6850243 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |