Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033330.3 | NP_001028502.1 | 9 - 10 | 1228 | 117 | |
NM_001290427.1 | NP_001277356.1 | 7 - 8 | 839 | 117 | ||
NM_001290428.1 | NP_001277357.1 | 6 - 7 | 772 | 117 | ||
GenBank mRNA | AK147370.1 | - | 9 - 10 | 1229 | 117 | |
AK147440.1 | - | 6 - 7 | 773 | 117 | ||
BC139429.1 | - | 7 - 8 | 684 | 8862451 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |