Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026149.4 | NP_080425.3 | 5 - 6 | 1344 | 106 | |
GenBank mRNA | AK014996.2 | - | 5 - 6 | 1344 | 106 | |
AK167855.1 | - | 5 - 6 | 876 | 106 | ||
AK168122.1 | - | 5 - 6 | 876 | 106 | ||
BC031583.1 | - | 5 - 6 | 910 | 4952090 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |