Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rims2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256382.1 | NP_001243311.1 | 24 - 25 | 4335 | 98 | |
NM_001256383.1 | NP_001243312.1 | 23 - 24 | 4269 | 98 | ||
NM_001256384.1 | NP_001243313.1 | 3 - 4 | 671 | 98 | ||
NM_053271.2 | NP_444501.1 | 24 - 25 | 4209 | 98 | ||
GenBank mRNA | AB021131.1 | - | 24 - 25 | 4163 | 98 | |
AK032619.1 | - | 3 - 4 | 617 | 98 | ||
AK129206.1 | - | 21 - 22 | 3433 | 98 | ||
BC118979.2 | - | 23 - 24 | 4217 | 40098740 | 98 | |
HM015529.1 | - | 24 - 25 | 4258 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |