Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166406.1 | NP_001159878.1 | 14 - 15 | 2069 | 75 | |
NM_001166407.1 | NP_001159879.1 | 14 - 15 | 2067 | 75 | ||
NM_009004.4 | NP_033030.1 | 14 - 15 | 1963 | 75 | ||
GenBank mRNA | AK051201.1 | - | 14 - 15 | 1881 | 75 | |
AK051995.1 | - | 14 - 15 | 1918 | 75 | ||
AK053280.1 | - | 14 - 15 | 1882 | 75 | ||
AK083412.1 | - | 14 - 15 | 1923 | 75 | ||
AK083697.1 | - | 14 - 15 | 1884 | 75 | ||
AK084732.1 | - | 14 - 15 | 1905 | 75 | ||
AK142764.1 | - | 14 - 15 | 1911 | 75 | ||
AK144844.1 | - | 14 - 15 | 1881 | 75 | ||
BC060608.1 | - | 14 - 15 | 2069 | 5720408 | 75 | |
Y09632.1 | - | 14 - 15 | 2118 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |