Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160404.1 | NP_001153876.1 | 7 - 8 | 1104 | 67 | |
NM_013814.3 | NP_038842.3 | 7 - 8 | 1192 | 67 | ||
GenBank mRNA | AK083391.1 | - | 7 - 8 | 1108 | 67 | |
AK148550.1 | - | 1 - 2 | 172 | 67 | ||
BC090962.1 | - | 6 - 7 | 982 | 30626890 | 67 | |
U73820.1 | - | 5 - 6 | 857 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |