Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177707.3 | NP_808375.1 | 6 - 7 | 960 | 74 | |
GenBank mRNA | AK036516.1 | - | 6 - 7 | 961 | 74 | |
BC067208.1 | - | 6 - 7 | 740 | 30286602 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |