Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012042.3 | NP_036172.1 | 14 - 15 | 1915 | 138 | |
GenBank mRNA | AF136441.1 | - | 13 - 14 | 1770 | 138 | |
AF176910.1 | - | 14 - 15 | 1804 | 138 | ||
AK147004.1 | - | 14 - 15 | 1897 | 138 | ||
AK164274.1 | - | 14 - 15 | 1882 | 138 | ||
AK165978.1 | - | 14 - 15 | 1856 | 138 | ||
BC004836.1 | - | 13 - 14 | 1724 | 138 | ||
BC029260.1 | - | 14 - 15 | 1915 | 2812400 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |