Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wibg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170869.2 | NP_001164340.1 | 1 - 2 | 108 | 61 | |
GenBank mRNA | BC049647.1 | - | 1 - 2 | 41 | 6707378 | 61 |
BI989958.1 | - | 1 - 2 | 108 | 61 | ||
BQ177652.1 | - | 1 - 2 | 34 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |