Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016756.4 | NP_058036.1 | 1 - 2 | 335 | 77 | |
NM_183417.3 | NP_904326.1 | 1 - 2 | 335 | 77 | ||
GenBank mRNA | AJ223732.1 | - | 1 - 2 | 237 | 77 | |
AK146845.1 | - | 1 - 2 | 315 | 77 | ||
AK148017.1 | - | 1 - 2 | 274 | 77 | ||
AK151979.1 | - | 1 - 2 | 285 | 77 | ||
AK152922.1 | - | 1 - 2 | 288 | 77 | ||
AK153983.1 | - | 1 - 2 | 287 | 77 | ||
AK154997.1 | - | 1 - 2 | 287 | 77 | ||
AK187629.1 | - | 1 - 2 | 71 | 77 | ||
AK189612.1 | - | 1 - 2 | 71 | 77 | ||
BC005654.1 | - | 1 - 2 | 296 | 3709730 | 77 | |
CN705407.1 | - | 1 - 2 | 335 | 77 | ||
U63337.1 | - | 1 - 2 | 263 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |