Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010296.2 | NP_034426.2 | 4 - 5 | 351 | 99 | |
GenBank mRNA | AB025922.1 | - | 4 - 5 | 361 | 99 | |
AB232673.1 | - | 3 - 4 | 282 | 99 | ||
AB232674.1 | - | 4 - 5 | 378 | 99 | ||
AB232675.1 | - | 4 - 5 | 511 | 99 | ||
AB232676.1 | - | 1 - 2 | 40 | 99 | ||
AB234616.1 | - | 5 - 6 | 862 | 99 | ||
AB234617.1 | - | 4 - 5 | 743 | 99 | ||
AF026305.1 | - | 4 - 5 | 351 | 99 | ||
CJ062804.1 | - | 3 - 4 | 335 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Inventoried Probe spans exons |