Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010296.2 | NP_034426.2 | 8 - 9 | 933 | 68 | |
GenBank mRNA | AB025922.1 | - | 8 - 9 | 943 | 68 | |
AF026305.1 | - | 8 - 9 | 933 | 68 | ||
BC131650.1 | - | 7 - 8 | 911 | 40110429 | 68 | |
CV782971.1 | - | 3 - 4 | 327 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |