Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019963.1 | NP_064347.1 | 19 - 20 | 1876 | 60 | |
GenBank mRNA | AF088862.1 | - | 19 - 20 | 1749 | 60 | |
AF187231.1 | - | 19 - 20 | 1873 | 60 | ||
AF206162.1 | - | 19 - 20 | 1876 | 60 | ||
AK030903.1 | - | 12 - 13 | 968 | 60 | ||
AK149924.1 | - | 19 - 20 | 1889 | 60 | ||
AK150070.1 | - | 19 - 20 | 1888 | 60 | ||
AK153405.1 | - | 17 - 18 | 1523 | 60 | ||
BC064827.1 | - | 8 - 9 | 873 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |