Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500019117.mm.1 |
TC0500002095.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500019117.mm.1 |
NM_001170555.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500019117.mm.1 |
NM_001170555.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500019117.mm.1 |
NM_001170556.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500019117.mm.1 |
NM_001170556.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500019117.mm.1 |
NM_145401.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500019117.mm.1 |
NM_145401.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500019115.mm.1 |
TC0500002095.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500019115.mm.1 |
NM_001170555.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500019115.mm.1 |
NM_001170555.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500019115.mm.1 |
NM_001170556.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500019115.mm.1 |
NM_001170556.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500019115.mm.1 |
NM_145401.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500019115.mm.1 |
NM_145401.2 |
Refseq |