Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmsb4x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021278.2 | NP_067253.1 | 1 - 2 | 112 | 64 | |
GenBank mRNA | AK010659.1 | - | 1 - 2 | 56 | 64 | |
AK088783.1 | - | 1 - 2 | 56 | 64 | ||
AK162286.1 | - | 1 - 2 | 57 | 64 | ||
AK169108.1 | - | 1 - 2 | 59 | 64 | ||
AK169718.1 | - | 1 - 2 | 56 | 64 | ||
BQ127748.1 | - | 1 - 2 | 112 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |