Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286607.1 | NP_001273536.1 | 8 - 9 | 1354 | 76 | |
NM_053242.4 | NP_444472.2 | 10 - 11 | 1754 | 76 | ||
NM_212435.1 | NP_997600.1 | 8 - 9 | 1417 | 76 | ||
GenBank mRNA | AF339106.1 | - | 7 - 8 | 1095 | 76 | |
AK051668.1 | - | 10 - 11 | 1737 | 76 | ||
AK082361.1 | - | 10 - 11 | 1426 | 76 | ||
AY079003.1 | - | 10 - 11 | 1383 | 76 | ||
BC058960.1 | - | 8 - 9 | 1351 | 6811877 | 76 | |
BC062926.1 | - | 8 - 9 | 1417 | 6851153 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |