Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201243.1 | NP_957695.1 | 1 | 1071 | 64 | |
GenBank mRNA | AY013766.1 | - | 1 | 1071 | 64 | |
BC139424.1 | - | 1 | 1167 | 8862446 | 64 | |
BC139426.1 | - | 1 | 1167 | 8862448 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |