Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111043.1 | NP_001104513.1 | 2 - 3 | 233 | 72 | |
GenBank mRNA | AK077992.1 | - | 2 - 3 | 109 | 72 | |
AK219072.1 | - | 1 - 2 | 54 | 72 | ||
BC085143.1 | - | 2 - 3 | 106 | 6415383 | 72 | |
J05609.1 | - | 2 - 3 | 107 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |