Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007664.4 | NP_031690.3 | 5 - 6 | 1354 | 71 | |
GenBank mRNA | AB008811.1 | - | 5 - 6 | 1031 | 71 | |
AK032826.1 | - | 5 - 6 | 794 | 71 | ||
AK146981.1 | - | 5 - 6 | 1551 | 71 | ||
BC022107.1 | - | 5 - 6 | 785 | 5100313 | 71 | |
M31131.1 | - | 5 - 6 | 1031 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |