Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012055.3 | NP_036185.1 | 1 - 2 | 79 | 58 | |
GenBank mRNA | AK160459.1 | - | 1 - 2 | 94 | 58 | |
AK167524.1 | - | 1 - 2 | 59 | 58 | ||
AK167933.1 | - | 1 - 2 | 78 | 58 | ||
BC005552.1 | - | 1 - 2 | 77 | 3585775 | 58 | |
BC050123.1 | - | 2407 | 58 | |||
U38940.1 | - | 1 - 2 | 80 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |