Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026458.4 | NP_080734.3 | 5 - 6 | 752 | 74 | |
GenBank mRNA | AK029665.1 | - | 5 - 6 | 756 | 74 | |
AY243470.1 | - | 5 - 6 | 752 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Recommended Assay Mus musculus Probe spans exons |