Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278943.1 | NP_001265872.1 | 13 - 14 | 2023 | 127 | |
NM_001278944.1 | NP_001265873.1 | 16 - 17 | 2492 | 127 | ||
GenBank mRNA | AY243472.1 | - | 16 - 17 | 2491 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Mus musculus Probe spans exons |