Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700038554.mm.1 |
NM_001122736.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038554.mm.1 |
NM_001122737.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038554.mm.1 |
NM_010514.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038554.mm.1 |
NM_010514.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038554.mm.1 |
TC0700004571.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700038556.mm.1 |
NM_001122737.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038556.mm.1 |
TC0700004571.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700038534.mm.1 |
NM_001122736.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038534.mm.1 |
NM_001122737.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038534.mm.1 |
NM_010514.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038534.mm.1 |
NM_010514.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038534.mm.1 |
TC0700004571.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700038535.mm.1 |
NM_001122736.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038535.mm.1 |
NM_001122737.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038535.mm.1 |
NM_010514.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038535.mm.1 |
NM_010514.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038535.mm.1 |
TC0700004571.mm.1 |
MTA TC |