Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700036872.mm.1 |
NM_001170744.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036872.mm.1 |
NM_001170744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036872.mm.1 |
TC0700004667.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036865.mm.1 |
NM_001170744.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036865.mm.1 |
NM_001170744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036865.mm.1 |
TC0700004667.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036879.mm.1 |
TC0700004667.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036874.mm.1 |
NM_001170744.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036874.mm.1 |
NM_001170744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036874.mm.1 |
TC0700004667.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036875.mm.1 |
NM_001170744.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036875.mm.1 |
NM_001170744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036875.mm.1 |
TC0700004667.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036869.mm.1 |
NM_001170744.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036869.mm.1 |
NM_001170744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036869.mm.1 |
TC0700004667.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036873.mm.1 |
NM_001170744.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036873.mm.1 |
NM_001170744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036873.mm.1 |
TC0700004667.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036870.mm.1 |
NM_001170744.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036870.mm.1 |
NM_001170744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036870.mm.1 |
TC0700004667.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036877.mm.1 |
TC0700004667.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036878.mm.1 |
TC0700004667.mm.1 |
MTA TC |