Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cthrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026778.2 | NP_081054.1 | 1 - 2 | 220 | 84 | |
GenBank mRNA | AK076498.1 | - | 1 - 2 | 223 | 84 | |
AK151282.1 | - | 1 - 2 | 221 | 84 | ||
BC131907.1 | - | 1 - 2 | 196 | 40130184 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |