Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30151915 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30151912 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30151913 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P453785 |
AK151282.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P453785 |
NM_026778.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P453785 |
AK003674.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P453785 |
AK076498.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P453785 |
AK151282.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P453785 |
NM_026778.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P453785 |
AK003674.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P453785 |
AK076498.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
376531 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
200667 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
234649 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
858145 |
AK080681.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
858145 |
AK168592.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
858145 |
BC016216.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
858145 |
BC025115.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
858145 |
BC078460.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
858145 |
NM_001110254.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
858145 |
NM_177358.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
737359 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
779467 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
144722 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P158432 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P150796 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P453785 |
AK151282.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P453785 |
NM_026778.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P453785 |
AK003674.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P453785 |
AK076498.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P453785 |
AK151282.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P453785 |
NM_026778.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P453785 |
AK003674.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P453785 |
AK076498.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P211702 |
AK090258.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P158432 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P150796 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P211702 |
AK090258.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P453785 |
AK151282.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P453785 |
NM_026778.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P453785 |
AK003674.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P453785 |
AK076498.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P453785 |
AK151282.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P453785 |
NM_026778.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P453785 |
AK003674.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P453785 |
AK076498.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P150796 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P158432 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30151913 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30151915 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30151912 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30152021 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151911 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151940 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151945 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30152016 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151935 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151929 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151942 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151950 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151918 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151978 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151936 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151967 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151927 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151917 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151932 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30152014 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151964 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151910 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151937 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151954 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151914 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151921 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151944 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151919 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151934 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151955 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151959 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151968 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151925 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30152018 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151939 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30152015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151923 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151915 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151920 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151933 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151960 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30151976 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151925 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12311680 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12238702 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151910 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151907 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151928 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12076900 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151940 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151944 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12311678 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151947 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12315651 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151935 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151927 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151939 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151911 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151949 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151921 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12465891 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151923 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151920 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12007162 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151937 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151950 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151919 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151945 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151918 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151934 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151932 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12315650 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30151936 |
- |
- |